Dicloxacillin sodium salt monohydrate
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-(2,6-Dichlorophenyl)-5-methyl-4-isoxazolyl penicillin sodium salt monohydrate, Sodium (2S,5R,6R)-6-(3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate, Dicloxacillin Sodium hydrate
CAS No.: 13412-64-1
Molecular Formula: C19H16Cl2N3NaO5S
Formula Weight: 492.33
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 222-225°
Solubility: Soluble in water
MDL No.: MFCD00056865
Applications: An antibiotic used in the treatment of staphyloccoci bacteria that are resistant to penicillin G
Formel:
C₁₉H₁₆Cl₂N₃NaO₅S·1H₂O Molecular Weight: 510.33 g/mol Schmelzpunkt: 222…225 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00056865 CAS-Nummer: 13412-64-1 EINECS: 603-794-2 Merck Index: 14,3084 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...