Emodin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,3,8-Trihydroxy-6-methylanthracene-9,10-dione, 1,3,8-Trihydroxy-6-methylanthraquinone
J61600.MC
J61600.MD
J61600.03
J61600.MCEA
119
EUR
J61600.MC
J61600.MD
J61600.03
Emodin
Emodin
CAS No.: 518-82-1
Molecular Formula: C15H10O5
Formula Weight: 270.24
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Orange
Beilstein Reference: 1888141
MDL No.: MFCD00001207
Applications: A protein tyrosine kinase inhibitor
Formel:
C₁₅H₁₀O₅ Molecular Weight: 270,24 g/mol |
MDL:
MFCD00001207 CAS-Nummer: 518-82-1 EINECS: 208-258-8 Merck Index: 14,3561 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...