Acarbose 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Prandase, Acarbose Hydrate, 4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose, Precose, (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4S,5S,6R)-3,4-Dihydroxy-6-methyl-5-(((1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl)amino)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, Glucobay

J61737.03 J61737.04 J61737.06
J61737.03EA 129 EUR
J61737.03 J61737.04 J61737.06
Acarbose 95%
Acarbose

Purity: 95% CAS No.: 56180-94-0 Molecular Formula: C25H43NO18 Formula Weight: 645.61 Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00869592


Applications: A reversible alpha-glucosidase inhibitor

Formel: C₂₅H₄₃NO₁₈
Molecular Weight: 645.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00869592
CAS-Nummer: 56180-94-0
EINECS: 260-030-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR