L-Tyrosin Dinatriumsalz-Dihydrat 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (S-3-(4-Hydroxyphenyl)alanine disodium salt dihydrate, Disodium 2-Amino-3-(4-oxidophenyl) propanoate dihydrate, L-3-(4-Hydroxyphenyl)alanine disodium salt dihydrate

J61770.22 J61770.36 J61770.A1
J61770.22EA 95.1 EUR
J61770.22 J61770.36 J61770.A1
L-Tyrosin Dinatriumsalz-Dihydrat 99%
L-Tyrosine disodium salt dihydrate

Purity: 99% CAS No.: 122666-87-9 Molecular Formula: C9H9NNa2O3.2H2O Formula Weight: 265.47 (225.15anhy) Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to pale brown Optical Rotation: -13° (c=4 in water) MDL No.: MFCD03840558

Formel: C₉H₉NO₃Na₂·2H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03840558
CAS-Nummer: 122666-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR