Spectinomycin dihydrochloride pentahydrate Reagenz für die Zellkultur

Lieferant: Thermo Fisher Scientific

Synonyme: (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate

J61820.06 J61820.14
J61820.06EA 104 EUR
J61820.06 J61820.14
Spectinomycin dihydrochloride pentahydrate Reagenz für die Zellkultur
Spectinomycin dihydrochloride pentahydrate

An aminocyclitol antibiotic that binds to the 30S ribosomal subunit of bacteria and interrupts protein synthesis


White powder.


Applications: An aminocyclitol antibiotic that binds to the 30S ribosomal subunit of bacteria and interrupts protein synthesis


Soluble in water

Formel: C₁₄H₂₄N₂O₇·2HCl·5H₂O
Lagertemperatur: Kühlschrank
MDL: MFCD00150886
CAS-Nummer: 22189-32-8
EINECS: 244-554-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR