Quinapril hydrochloride 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Quinapril HCl, (S)-2-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)-L-alanyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride
J61913.MC
J61913.03
J61913.06
J61913.MCEA
47.1
EUR
J61913.MC
J61913.03
J61913.06
Quinapril hydrochloride 98%
Quinapril hydrochloride
Purity: 98%
CAS No.: 82586-55-8
Molecular Formula: C25H30N2O5.HCl
Formula Weight: 474.99
Physical Form: Solid
Appearance: White
Melting Point: 120-130°
MDL No.: MFCD00889215
Applications: Angiotensin-converting enzyme inhibitor
Formel:
C₂₅H₃₀N₂O₅·HCl Molecular Weight: 474,99 g/mol Schmelzpunkt: 120…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00889215 CAS-Nummer: 82586-55-8 Merck Index: 14,8051 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...