Metoprolol-Tartrat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-(Isopropylamino)-3-[4-(beta-methoxyethyl)phenoxy]-2-propanol (+)-Tartrate
J61920.06
J61920.14
J61920.06EA
77.2
EUR
J61920.06
J61920.14
Metoprolol-Tartrat ≥98%
Metoprolol-Tartrat
Purity: 98+%
CAS No.: 56392-17-7
Molecular Formula: (C15H25NO3)2.C4H6O6
Formula Weight: 684.82
Physical Form: Powder
Appearance: White
Melting Point: 120°
MDL No.: MFCD00056257
Applications: A beta-adrenergic blocker
Formel:
C₁₅H₂₅NO₃ Molecular Weight: 342.41 g/mol Schmelzpunkt: 120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00056257 CAS-Nummer: 56392-17-7 EINECS: 260-148-9 Merck Index: 14,6151 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...