Inosine 5'-monophosphate disodium salt hydrate
Lieferant: Thermo Scientific
Synonyme:
5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate
J61959.06
J61959.14
J61959.22
J61959.06EA
32.7
EUR
J61959.06
J61959.14
J61959.22
Inosine 5'-monophosphate disodium salt hydrate
Inosine 5'-monophosphate disodium salt hydrate
CAS No.: 4691-65-0
Molecular Formula: C10H11N4Na2O8P.xH2O
Formula Weight: 392.17(anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
MDL No.: MFCD00036201
Formel:
C₁₀H₁₁N₄Na₂O₈P·xH₂O Molecular Weight: 392,17 g/mol (anhy) |
MDL:
MFCD00150372 CAS-Nummer: 4691-65-0 EINECS: 225-146-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...