5-Aminofluorescein (isomer I) 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one (isomer I), 5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-Aminofluorescein, Fluoresceinamine, isomer I, 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, Fluoresceinamine (isomer I)

J62175.03 J62175.06
J62175.03EA 126 EUR
J62175.03 J62175.06
5-Aminofluorescein (isomer I) 96%
5-Aminofluorescein (isomer I)

Purity: 96% CAS No.: 3326-34-9 Molecular Formula: C20H13NO5 Formula Weight: 347.32 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Dark red to brown Melting Point: 223° dec. Beilstein Reference: 48395 MDL No.: MFCD00005052


Applications: A fluorescent labeling reagent

Formel: C₂₀H₁₃NO₅
Molecular Weight: 347,33 g/mol
Schmelzpunkt: 223 °C (Zersetzung)
MDL: MFCD00005052
CAS-Nummer: 3326-34-9
EINECS: 222-043-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR