Pirenzepine dihydrochloride 99%
Lieferant: Thermo Scientific
Synonyme:
11-(2-(4-Methylpiperazin-1-yl)acetyl)-5,11-dihydro-6H-benzo[e]pyrido[3,2-b][1,4]diazepin-6-one dihydrochloride
J62252.MC
J62252.03
J62252.MCEA
39.4
EUR
J62252.MC
J62252.03
Pirenzepine dihydrochloride 99%
Pirenzepine dihydrochloride
Purity: 99%
CAS No.: 29868-97-1
Molecular Formula: C19H21N5O2.2HCl
Formula Weight: 424.32
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 192-198°
MDL No.: MFCD00055214
Applications: A M1 muscarinic receptor antagonist
Formel:
C₁₉H₂₁N₅O₂·2HCl Molecular Weight: 424,32 g/mol Schmelzpunkt: 192…198 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055214 CAS-Nummer: 29868-97-1 EINECS: 249-907-5 Merck Index: 14,7491 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...