L-Cystin Dihydrochlorid 99%
Lieferant: Thermo Scientific
Synonyme:
(2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid) dihydrochloride, H-Cystine-OH·2HCl
J62292.14
J62292.22
J62292.30
J62292.14EA
68.5
EUR
J62292.14
J62292.22
J62292.30
L-Cystin Dihydrochlorid 99%
L-Cystin Dihydrochlorid
Purity: 99%
CAS No.: 30925-07-6
Molecular Formula: C6H12N2O4S2.2HCl
Formula Weight: 313.22
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 228-232°
MDL No.: MFCD00070399
Formel:
C₆H₁₂N₂O₄S₂·2HCl Molecular Weight: 313,23 g/mol Schmelzpunkt: 228…232 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070399 CAS-Nummer: 30925-07-6 EINECS: 250-391-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...