β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)) 97%
Lieferant: Thermo Scientific
Synonyme:
NAD+, Nicotinamid-Adenin-Dinucleotid, oxidiert, Nadid, Nicotinsäureamid-Adenin-Dinucleotid, β-Nicotinamid-Adenin-Dinucleotid
J62337.03
J62337.06
J62337.14
J62337.03EA
51.2
EUR
J62337.03
J62337.06
J62337.14
β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)) 97%
β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)
Purity: 98+%
CAS No.: 53-84-9
Molecular Formula: C21H27N7O14P2
Formula Weight: 663.43
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water at 50mg/ml
Beilstein Reference: 3584133
MDL No.: MFCD00036253
Formel:
C₂₁H₂₇N₇O₁₄P₂ Molecular Weight: 663,43 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00036253 CAS-Nummer: 53-84-9 EINECS: 200-184-4 Merck Index: 14,6344 |