Penicillin V potassium salt

Lieferant: Thermo Scientific
Gefahr

Synonyme: Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin V Potassium

J62442.09 J62442.14 J62442.22
J62442.09EA 55.2 EUR
J62442.09 J62442.14 J62442.22
Penicillin V potassium salt
Penicillin V potassium salt

Inhibits bacterial cell wall synthesis.


White to off-white powder.


Applications: Inhibits bacterial cell wall synthesis

Formel: C₁₆H₁₇KN₂O₅S
Molecular Weight: 388,48 g/mol
Schmelzpunkt: 197…202 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00051771
CAS-Nummer: 132-98-9
EINECS: 205-086-5
Merck Index: 14,7100

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR