Rapamycin ≥99%
Lieferant: Thermo Scientific
Synonyme:
(3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone
Purity: 99+%
CAS No.: 53123-88-9
Molecular Formula: C51H79NO13
Formula Weight: 914.17
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
Solubility: Soluble in DMSO at 50mg/ml or methanol at 25mg/ml
MDL No.: MFCD00867594
Applications: Blocks cytokine-mediated signal transduction pathways
Formel:
C₅₁H₇₉NO₁₃ Molecular Weight: 914,19 g/mol |
MDL:
MFCD00867594 CAS-Nummer: 53123-88-9 Merck Index: 14,8114 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...