(S)-(+)-Camptothecin ≥90%
Lieferant: Thermo Scientific
Gefahr
Synonyme: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
J62523.MD J62523.03 J62523.06
J62523.MDEA 87.2 EUR
J62523.MD J62523.03 J62523.06
(S)-(+)-Camptothecin ≥90%
(S)-(+)-Camptothecin
Cytotoxic antitumor agent that inhibits topoisomerase I.
Formula Weight: 348.35 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Yellow Melting Point: 260° dec. Beilstein Reference: 631069 UN No.: UN1544 MDL No.: MFCD00081076
Applications: Cytotoxic antitumor agent that inhibits topoisomerase I
Formel: C₂₀H₁₆N₂O₄
Molecular Weight: 348,36 g/mol
Schmelzpunkt: 260 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD00081076
CAS-Nummer: 7689-03-4
UN: 1544
ADR: 6.1,III
Merck Index: 14,1735