Flumazenil 98%
Lieferant: Thermo Scientific
Synonyme:
Ethyl 8-fluoro-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylate
J62537.MC
J62537.ME
J62537.MCEA
416
EUR
J62537.MC
J62537.ME
Flumazenil 98%
Flumazenil
Purity: 98%
CAS No.: 78755-81-4
Molecular Formula: C15H14FN3O3
Formula Weight: 303.29
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in DMSO to 25mM
MDL No.: MFCD00242764
Applications: Benzodiazepine antagonist
Formel:
C₁₅H₁₄FN₃O₃ Molecular Weight: 303,29 g/mol |
MDL:
MFCD00242764 CAS-Nummer: 78755-81-4 Merck Index: 14,4135 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...