Pyridoxamine dihydrochloride Reagenz für die Zellkultur
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride
J62679.27
J62679.06
J62679.09
J62679.27EA
108
EUR
J62679.27
J62679.06
J62679.09
Pyridoxamine dihydrochloride Reagenz für die Zellkultur
Pyridoxamine dihydrochloride
CAS No.: 524-36-7
Molecular Formula: C8H12N2O2.2HCl
Formula Weight: 241.12
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Off-white
Melting Point: 220-224°
Solubility: Soluble in DMSO, water, or methanol
Beilstein Reference: 3632748
MDL No.: MFCD00012808
Applications: A vitamin in the Vitamin B-6 family
Formel:
C₈H₁₂N₂O₂·2HCl Molecular Weight: 241.12 g/mol Schmelzpunkt: 220…224 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00012808 CAS-Nummer: 524-36-7 EINECS: 208-357-6 Merck Index: 14,7980 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...