Nifedipin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Adalat, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Dimethyl Ester

J62811.06 J62811.14 J62811.22
J62811.06EA 63.7 EUR
J62811.06 J62811.14 J62811.22
Nifedipin 98%
Nifedipin

Purity: 98% CAS No.: 21829-25-4 Molecular Formula: C17H18N2O6 Formula Weight: 346.33 Physical Form: Powder Appearance: Off-white to yellow Melting Point: 171-175° MDL No.: MFCD00057326


Applications: L-type calcium channel blocker; induces apoptosis in human glioblastoma cells

Formel: C₁₇H₁₈N₂O₆
Molecular Weight: 346,34 g/mol
Schmelzpunkt: 171…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00057326
CAS-Nummer: 21829-25-4
EINECS: 244-598-3
Merck Index: 14,6528

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR