Candesartan 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-((2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylic acid, 1-2-1H-Tetrazol-5-yl-1-1-biphenyl-4-yl-methyl-2-ethoxy-1H-benzo-d-imidazole-7-carboxylic-acid, 1-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylic acid
J62818.MC
J62818.03
J62818.MCEA
63.3
EUR
J62818.MC
J62818.03
Candesartan 98%
Candesartan
Purity: 98%
CAS No.: 139481-59-7
Molecular Formula: C24H20N6O3
Formula Weight: 440.45
Physical Form: Powder
Appearance: White
Melting Point: 183-185°
MDL No.: MFCD00081076
Applications: Angiotensin II receptor antagonist
Formel:
C₂₄H₂₀N₆O₃ Molecular Weight: 440,45 g/mol Schmelzpunkt: 183…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081076 CAS-Nummer: 139481-59-7 Merck Index: 14,1739 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...