Cimetidin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Cyano-1-methyl-3-[2-(5-methyl-1H-imidazol-4-yl-methylthio)ethyl]guanidine, N-Cyano-N'-methyl-N''-[2-[[(4-methyl-1H-imidazol-5-yl)methyl]thio]ethyl]guanidine

J62825.06 J62825.09 J62825.14
J62825.06EA 81.3 EUR
J62825.06 J62825.09 J62825.14
Cimetidin ≥98%
Cimetidine

Purity: 98+% CAS No.: 51481-61-9 Molecular Formula: C10H16N6S Formula Weight: 252.34 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Melting Point: 139-144° MDL No.: MFCD00133296


Applications: H2 histamine receptor antagonist

Formel: C₁₀H₁₆N₆S
Schmelzpunkt: 139…144 °C
MDL: MFCD00133296
CAS-Nummer: 51481-61-9
EINECS: 257-232-2
Merck Index: 14,2279

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR