Chelerythrine chloride ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
J62906.MA
J62906.MB
J62906.MAEA
218
EUR
J62906.MA
J62906.MB
Chelerythrine chloride ≥99%
Chelerythrine chloride
Purity: 99+%
CAS No.: 3895-92-9
Molecular Formula: C21H18ClNO4
Formula Weight: 383.82
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Yellow to orange
Solubility: Soluble in DMSO at 2mg/ml
MDL No.: MFCD00060717
Applications: A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells
Formel:
C₂₁H₁₈ClNO₄ |
MDL:
MFCD00060717 CAS-Nummer: 3895-92-9 EINECS: 223-444-9 Merck Index: 14,2051 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...