Orlistat 98%
Lieferant: Thermo Scientific
Synonyme:
N-Formyl-L-leucine (1S)-1-[[(2S,3S)-3-Hexyl-4-oxo-2-oxetanyl]methyl]dodecyl Ester, (S)-(S)-1-((2S,3S)-3-Hexyl-4-oxooxetan-2-yl)tridecan-2-yl 2-formamido-4-methylpentanoate
J62999.MF
J62999.MC
J62999.MD
J62999.MFEA
69.2
EUR
J62999.MF
J62999.MC
J62999.MD
Orlistat 98%
Orlistat
Purity: 98%
CAS No.: 96829-58-2
Molecular Formula: C29H53NO5
Formula Weight: 495.73
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
Melting Point:
Applications: Novel inhibitor of fatty acid sythase. Halts tumor cell proliferation and induces tumor cell apoptosis
Formel:
C₂₉H₅₃NO₅ Molecular Weight: 495,75 g/mol Schmelzpunkt: <50 °C |
MDL:
MFCD05662360 CAS-Nummer: 96829-58-2 Merck Index: 14,6869 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...