Trimethoprim
Lieferant: Thermo Scientific
Synonyme:
5-(3,4,5-Trimethoxybenzyl)pyrimidine-2,4-diamine, 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine
CAS No.: 738-70-5
Molecular Formula: C14H18N4O3
Formula Weight: 290.32
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 199-203°
Solubility: Trimethoprim is soluble in water at 0.4 mg/ml. It is alsosoluble in DMSO at 50 mg/ml and chloroform at 18.2 mg/ml.
Beilstein Reference: 625127
MDL No.: MFCD00036761
Applications: An antibacterial and inhibitor of formylation. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme
Formel:
C₁₄H₁₈N₄O₃ Molecular Weight: 290,32 g/mol Schmelzpunkt: 199…203 °C |
MDL:
MFCD00036761 CAS-Nummer: 738-70-5 EINECS: 212-006-2 Merck Index: 14,9709 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...