Capsazepine ≥99%

Lieferant: Thermo Scientific

Synonyme: N-(4-Chlorophenethyl)-7,8-dihydroxy-4,5-dihydro-1H-benzo[c]azepine-2(3H)-carbothioamide

J63055.MA J63055.MF
J63055.MAEA 233 EUR
J63055.MA J63055.MF
Capsazepine ≥99%
Capsazepine

Purity: 99+% CAS No.: 138977-28-3 Molecular Formula: C19H21ClN2O2S Formula Weight: 376.90 Physical Form: Powder Appearance: White to off-white Melting Point: 159-161° Solubility: Soluble in ethanol or DMSO MDL No.: MFCD00153778


Applications: A competitive antagonist for capsaicin and resiniferatoxin

Formel: C₁₉H₂₁ClN₂O₂S
Molecular Weight: 376,90 g/mol
Schmelzpunkt: 159…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00153778
CAS-Nummer: 138977-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR