Trazodone hydrochloride
Lieferant: Thermo Scientific
|
Achtung
|
J63070.03
J63070.06
J63070.03EA
75.1
EUR
J63070.03
J63070.06
Trazodone hydrochloride
Trazodone hydrochloride
CAS No.: 25332-39-2
Molecular Formula: C19H22ClN5O.HCl
Formula Weight: 408.33
Physical Form: Powder
Appearance: Off-white
Melting Point: 222-227°
Solubility: Soluble at 25mg/ml in methanol. Also soluble in 0.1M HCl or DMSO. Insoluble in water
MDL No.: MFCD00079603
Applications: A serotonin uptake inhibitor
Formel:
C₁₉H₂₃Cl₂N₅O Schmelzpunkt: 222…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079603 CAS-Nummer: 25332-39-2 EINECS: 246-855-5 Merck Index: 14,9579 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...