L-Cystine disodium salt monohydrate ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (H-Cys-OH)2.2Na.H2O

J63071.18 J63071.30
J63071.18EA 58.6 EUR
J63071.18 J63071.30
L-Cystine disodium salt monohydrate ≥98%
L-Cystine disodium salt monohydrate

Purity: 98+% CAS No.: 199329-53-8 Molecular Formula: C6H10N2Na2O4S2.H2O Formula Weight: 302.27 (284.26anhy) Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00243291

Formel: C₆H₁₀N₂Na₂O₄S₂·H₂O
Molecular Weight: 302,27 g/mol (284,26 anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00243291
CAS-Nummer: 199329-53-8
EINECS: 265-025-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR