Ornidazole 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(3-Chloro-2-hydroxypropyl)-2-methyl-5-nitroimidazole, 1-Chloro-3-(2-methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol

J63100.06 J63100.14
J63100.06EA 60 EUR
J63100.06 J63100.14
Ornidazole 99%
Ornidazole

Purity: 99% CAS No.: 16773-42-5 Molecular Formula: C7H10ClN3O3 Formula Weight: 219.63 Physical Form: Powder Appearance: White to pale yellow Melting Point: 84-86° Solubility: Soluble in ethanol at 50mg/ml MDL No.: MFCD00057960


Applications: Interferes with electron transport in metabolic pathways

Formel: C₇H₁₀ClN₃O₃
Molecular Weight: 219.63 g/mol
Schmelzpunkt: 84…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00057960
CAS-Nummer: 16773-42-5
EINECS: 240-826-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR