Ponceau S 0,2% (v/v) in 5% Essigsäure
Lieferant: Thermo Scientific
Synonyme:
3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
J63139.AP
J63139.K2
J63139.APEA
83.1
EUR
J63139.AP
J63139.K2
Ponceau S 0,2% (v/v) in 5% Essigsäure
Ponceau S
CAS No.: 6226-79-5
Molecular Formula: C22H12N4Na4O13S4
Formula Weight: 760.56
Physical Form: Liquid
Appearance: Red
MDL No.: MFCD00003892
Formel:
C₂₂H₁₂N₄Na₄O₁₃S₄ Molecular Weight: 760,56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003892 CAS-Nummer: 6226-79-5 |