Naringin

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone 7-rhamnoglucoside, Naringenin 7-rhamnoglucoside

J63166.14 J63166.22
J63166.14EA 47.6 EUR
J63166.14 J63166.22
Naringin
Naringin

CAS No.: 10236-47-2 Molecular Formula: C27H32O14 Formula Weight: 580.53 Physical Form: Powder Appearance: White to pale yellow Beilstein Reference: 102012 MDL No.: MFCD00148888


Applications: A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activity

Formel: C₂₇H₃₂O₁₄
Molecular Weight: 580,53 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00148888
CAS-Nummer: 10236-47-2
EINECS: 233-566-4
Merck Index: 14,6425

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR