Econazolnitrat

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-[2-(4-Chlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole Nitrate, Ecostatin, 1-[2-(4-Chlorophenylmethoxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole nitrate

J63173.06 J63173.14
J63173.06EA 69.1 EUR
J63173.06 J63173.14
Econazolnitrat
Econazole nitrate

CAS No.: 24169-02-6 Molecular Formula: C18H15Cl3N2O.HNO3 Formula Weight: 444.70 Physical Form: Powder Appearance: White to off-white Melting Point: 158-163° Solubility: Soluble at 0.4mg/ml in DMSO. Sparingly soluble in water. MDL No.: MFCD00058160

Formel: C₁₈H₁₅Cl₃N₂O·HNO₃
Molecular Weight: 444,69 g/mol
Schmelzpunkt: 158…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00058160
CAS-Nummer: 24169-02-6
EINECS: 246-053-5
Merck Index: 14,3502

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR