(+)-cis-Diltiazem hydrochloride

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2S,3S)-(+)-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, Diltiazem Hydrochloride, (+)-cis-Diltiazem HCl, (+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, (2S,3S)-5-(2-(Dimethylamino)ethyl)-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate hydrochloride

J63245.06 J63245.14
J63245.06EA 129 EUR
J63245.06 J63245.14
(+)-cis-Diltiazem hydrochloride
(+)-cis-Diltiazem hydrochloride

CAS No.: 33286-22-5 Molecular Formula: C22H26N2O4S.HCl Formula Weight: 450.98 Physical Form: Powder Appearance: White Melting Point: 207-212° Solubility: Soluble in water at 50mg/ml Beilstein Reference: 4228706 MDL No.: MFCD00069252


Applications: An L-type calcium channel blocker

Formel: C₂₂H₂₆N₂O₄S·HCl
Schmelzpunkt: 207…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00069252
CAS-Nummer: 33286-22-5
EINECS: 251-443-3
Merck Index: 14,3202

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR