3,3',5'-Triiodo-L-thyronine sodium salt
Lieferant: Thermo Fisher Scientific
Synonyme:
Liothyronine sodium salt, Sodium (S)-2-amino-3-(4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl)propanoate
J63312.ME
J63312.03
J63312.MEEA
112
EUR
J63312.ME
J63312.03
3,3',5'-Triiodo-L-thyronine sodium salt
3,3',5'-Triiodo-L-thyronine sodium salt
CAS No.: 55-06-1
Molecular Formula: C15H11I3NNaO4
Formula Weight: 672.96
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to pale brown
Melting Point: 205°
Beilstein Reference: 8179867
MDL No.: MFCD00002594
Formel:
C₁₅H₁₁I₃NNaO₄ Molecular Weight: 672.959 g/mol |
MDL:
MFCD00002594 CAS-Nummer: 55-06-1 EINECS: 200-223-5 |