Dipyridamol

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,6-Bis(diethanolamine)-4,8-dipiperidinopyrimido[5,4-d]pyrimidine, 2,2',2'',2'''-((4,8-Di(piperidin-1-yl)pyrimido[5,4-d]pyrimidine-2,6-diyl)bis(azanetriyl))tetrakis(ethan-1-ol), 2,2',2'',2'''-[(4,8-Dipiperidinylpyrimido[5,4-d]pyrimidine-2,6-diyl)dinitrilo]tetrakisethanol

J63329.06 J63329.14
J63329.06EA 49.9 EUR
J63329.06 J63329.14
Dipyridamol
Dipyridamole

CAS No.: 58-32-2 Molecular Formula: C24H40N8O4 Formula Weight: 504.63 Physical Form: Powder Appearance: Yellow Melting Point: 165-167° MDL No.: MFCD00010555


Applications: An inhibitor of adenosine transport and phosphodiesterase

Formel: C₂₄H₄₀N₈O₄
Molecular Weight: 504,64 g/mol
Schmelzpunkt: 165…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010555
CAS-Nummer: 58-32-2
EINECS: 200-374-7
Merck Index: 14,3346

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR