Cytidine-5'-monophosphate disodium salt ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate

J63376.03 J63376.06 J63376.14
J63376.03EA 34 EUR
J63376.03 J63376.06 J63376.14
Cytidine-5'-monophosphate disodium salt ≥99%
Cytidine-5'-monophosphate disodium salt

Purity: 99+% CAS No.: 6757-06-8 Molecular Formula: C9H12N3Na2O8P Formula Weight: 367.16 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00064355

Formel: C₉H₁₂N₃Na₂O₈P
Molecular Weight: 367.16 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00064355
CAS-Nummer: 6757-06-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR