Diatrizoesäure

Lieferant: Thermo Scientific

Synonyme: 3,5-Di(acetamido)-2,4,6-triiodobenzoic Acid, 3,5-Bis(acetamido)-2,4,6-triiodobenzoic Acid, 3,5-Diacetamido-2,4,6-triiodobenzoic acid

J63444.14 J63444.22
J63444.14EA 85.4 EUR
J63444.14 J63444.22
Diatrizoesäure
Diatrizoic acid

CAS No.: 117-96-4 Molecular Formula: C11H9I3N2O4 Formula Weight: 613.90 Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00069960


Applications: A high-osmolality radiocontrast agent

Formel: C₁₁H₉I₃N₂O₄
Molecular Weight: 613,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00069960
CAS-Nummer: 117-96-4
EINECS: 204-223-6
Merck Index: 14,2992

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR