Tamoxifen 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: cis-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene

J63509.ME J63509.03 J63509.06
J63509.MEEA 137 EUR
J63509.ME J63509.03 J63509.06
Tamoxifen 98%
Tamoxifen

Inhibitor of protein kinase C and induces apoptosis in cancer cells.


Tamoxifen is used in the treatment of both early and advanced estrogen receptor positive breast cancer in pre- and post-menopausal women. It is also used in the treatment of infertility in women with anovulatory disorders as well as to prevent estrogen-related gynecomastia. It is utilized in the study on lipopolysaccharide (LPS)-induced microglial activation to know its effect.


Applications: Inhibitor of protein kinase C and induces apoptosis in cancer cells

Formel: C₂₆H₂₉NO
Molecular Weight: 371,524 g/mol
MDL: MFCD00010454
CAS-Nummer: 10540-29-1
EINECS: 234-118-0
Merck Index: 14,9048

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR