Cetirizin-dihydrochlorid ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cetirizine 2HCl, 2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)acetic acid dihydrochloride
J63549.06
J63549.09
J63549.14
J63549.06EA
156
EUR
J63549.06
J63549.09
J63549.14
Cetirizin-dihydrochlorid ≥99%
Cetirizine dihydrochloride
Purity: 99+%
CAS No.: 83881-52-1
Molecular Formula: C21H25ClN2O3.2HCl
Formula Weight: 461.82
Physical Form: Solid
Appearance: White
MDL No.: MFCD00941428
Applications: Histamine H-1 receptor antagonist. Inhibits activation of eosinophils and neutrophils
Formel:
C₂₁H₂₅ClN₂O₃ Molecular Weight: 461,81 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00941428 CAS-Nummer: 83881-52-1 Merck Index: 14,2022 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...