Etoposid

Lieferant: Thermo Scientific
Gefahr

Synonyme: (5R,5aR,8aR,9S)-9-(((4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-methylhexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one, 4'-Demethylepipodophyllotoxin 9-(4,6-O-Ethylidene-beta-D-glucopyranoside)

J63651.MB J63651.MC
J63651.MBEA 78.7 EUR
J63651.MB J63651.MC
Etoposid
Etoposide

CAS No.: 33419-42-0 Molecular Formula: C29H32O13 Formula Weight: 588.56 Physical Form: Powder Appearance: White Melting Point: 236-251° Optical Rotation: -107° (c=1 in methanol/dichloromethane 1:9) MDL No.: MFCD00869325


Applications: Topoisomerase inhibitor and inducer of apoptosis

Formel: C₂₉H₃₂O₁₃
Molecular Weight: 588,56 g/mol
Schmelzpunkt: 236…251 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00869325
CAS-Nummer: 33419-42-0
EINECS: 251-509-1
Merck Index: 14,3886

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR