L-Cystin Dihydrochlorid 98% (durch titrimetrische Analyse an Trockenmasse)

Lieferant: Thermo Scientific

Synonyme: (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid) dihydrochloride, H-Cystine-OH·2HCl

J63717.30 J63717.A1
J63717.30EA 163 EUR
J63717.30 J63717.A1
L-Cystin Dihydrochlorid 98% (durch titrimetrische Analyse an Trockenmasse)
L-Cystin Dihydrochlorid

CAS No.: 30925-07-6 Molecular Formula: C6H12N2O4S2.2HCl Formula Weight: 313.22 Physical Form: Powder Appearance: White to pale yellow Melting Point: 228-232° MDL No.: MFCD00070399

Formel: C₆H₁₂N₂O₄S₂·2HCl
Molecular Weight: 313,23 g/mol
Schmelzpunkt: 228…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00070399
CAS-Nummer: 30925-07-6
EINECS: 250-391-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR