Gossypol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,2'-bis(8-Formyl-1,6,7-trihydroxy-5-isopropyl-3-methylnaphthalene), 1,1',6,6',7,7'-Hexahydroxy-5,5'-diisopropyl-3,3'-dimethyl-[2,2'-binaphthalene]-8,8'-dicarbaldehyde, Thespesin
J63767.MC
J63767.MD
J63767.MCEA
206
EUR
J63767.MC
J63767.MD
Gossypol ≥98%
Gossypol
Purity: 98+%
CAS No.: 303-45-7
Molecular Formula: C30H30O8
Formula Weight: 518.56
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Yellow to brown
MDL No.: MFCD00017352
Applications: Lipophilic agent derived from cottonseed
Formel:
C₃₀H₃₀O₈ |
MDL:
MFCD00017352 CAS-Nummer: 303-45-7 Merck Index: 14,4528 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...