Kaffeesäuremethylester
Lieferant: Thermo Scientific
Synonyme:
3,4-Dihydroxycinnamic Acid Methyl Ester, Methyl 3,4-Dihydroxycinnamate, Caffeic Acid Methyl Ester
J63786.MD
J63786.03
J63786.MDEA
86.5
EUR
J63786.MD
J63786.03
Kaffeesäuremethylester
Kaffeesäuremethylester
CAS No.: 3843-74-1
Molecular Formula: C10H10O4
Formula Weight: 194.19
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 158-161°
MDL No.: MFCD00210468
Applications: Inhibitor of protein tyrosine kinase
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194,19 g/mol Schmelzpunkt: 158…161 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00210468 CAS-Nummer: 3843-74-1 Merck Index: 14,1635 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...