Ethyl violet
Lieferant: Thermo Scientific
Synonyme:
Basic Violet 4, N-(4-(Bis(4-(diethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium chloride
J63951.14
J63951.22
J63951.14EA
40.7
EUR
J63951.14
J63951.22
Ethyl violet
Ethyl violet
CAS No.: 2390-59-2
Molecular Formula: C31H42ClN3
Formula Weight: 492.15
Physical Form: Powder
Appearance: Olive green
Beilstein Reference: 4117566
MDL No.: MFCD00011765
Formel:
C₃₁H₄₂ClN₃ Molecular Weight: 492,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011765 CAS-Nummer: 2390-59-2 EINECS: 219-231-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...