Ticlopidine hydrochloride
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Triclopidine hydrochloride, Ticlopidine HCl, 5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride
J63971.06
J63971.14
J63971.06EA
61.1
EUR
J63971.06
J63971.14
Ticlopidine hydrochloride
Ticlopidine hydrochloride
CAS No.: 53885-35-1
Molecular Formula: C14H14ClNS.HCl
Formula Weight: 300.25
Physical Form: Powder
Appearance: White
MDL No.: MFCD00079606
Applications: Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines
Formel:
C₁₄H₁₄ClNS Molecular Weight: 300,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079606 CAS-Nummer: 53885-35-1 EINECS: 258-837-4 Merck Index: 14,9429 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...