Safflower oil
Lieferant: Thermo Scientific
Synonyme: Carthamus tinctorius oil, Triglyceride of Fatty Acids, Fats and Glyceridic oils, safflower, Oils, glyceridic, safflower, Thistle oil, Safflower seed oil
J63982.AP J63982.K2
J63982.APEA 41 EUR
J63982.AP J63982.K2
Safflower oil
Safflower oil
CAS No.: 8001-23-8 Flash Point: 113°(235°F) Physical Form: Oil Density: 0.921 Refractive Index: 1.476 Appearance: Pale yellow MDL No.: MFCD00132216
Dichte: 0,921
Flammpunkt: >113 °C (235 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00132216
CAS-Nummer: 8001-23-8
EINECS: 232-276-5