Azadibenzocyclooctyne-NHS ester, with 4 carbon linker
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
ADIBO-NHS ester, with 4 carbon linker
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification.
- Moisture sensitive
It is used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.
Soluble in DMF, THF, DMSO and DCM .
Bitte beachten Moisture and heat sensitive. Store in cool place. Incompatible with strong oxidizng agents.