Cytosine-β-D-arabinofuranoside hydrochloride

Lieferant: Thermo Scientific
Achtung

Synonyme: Arabinosylcytosine hydrochloride, Cytarabine HCI, Cytarabine HCl, 4-Amino-1-((2S,3R,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride, 1-(β-D-Arabinofuranosyl)cytosine HCl

J65671.03 J65671.06 J65671.09
J65671.03EA 39.3 EUR
J65671.03 J65671.06 J65671.09
Cytosine-β-D-arabinofuranoside hydrochloride
Cytosine-beta-D-arabinofuranoside hydrochloride

CAS No.: 69-74-9 Molecular Formula: C9H13N3O5.HCl Formula Weight: 279.68 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 197-198° MDL No.: MFCD00012839


Applications: Interferes with the synthesis of DNA

Formel: C₉H₁₄ClN₃O₅
Molecular Weight: 279,677 g/mol
Schmelzpunkt: 197…198 °C
MDL: MFCD00012839
CAS-Nummer: 69-74-9
EINECS: 200-713-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR