2'-Desoxy-2'-fluoruridin 97%

Lieferant: Thermo Scientific

Synonyme: 2'-Fluoro-2'-deoxyuridine, 1-((2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

J65948.03 J65948.06 J65948.14
J65948.03EA 29.1 EUR
J65948.03 J65948.06 J65948.14
2'-Desoxy-2'-fluoruridin 97%
2'-Deoxy-2'-fluorouridine

Purity: 99% CAS No.: 784-71-4 Molecular Formula: C9H11FN2O5 Formula Weight: 246.19 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD01317293

Formel: C₉H₁₁FN₂O₅
Molecular Weight: 246,19 g/mol
MDL: MFCD01317293
CAS-Nummer: 784-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR