Clarithromycin 960 - 1040 µg/mg ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-(((2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-14-ethyl-12,13-dihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-7-methoxy-3,5,7,9,11,13-hexamethyloxacyclotetradecane-2,10-dione

J66005.MC J66005.03 J66005.06
J66005.MCEA 21.6 EUR
J66005.MC J66005.03 J66005.06
Clarithromycin 960 - 1040 µg/mg ≥97%
Clarithromycin

A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit.


White powder


Soluble in DMSO

Formel: C₃₈H₆₉NO₁₃
Molecular Weight: 747,96 g/mol
Siedepunkt: 86 °C
Schmelzpunkt: 217…220 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00865140
CAS-Nummer: 81103-11-9
Merck Index: 14,2339

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR