Sulphadoxin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: sulphadoxine, 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzenesulphonamide, 4-Amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulphonamide

J66268.06 J66268.14 J66268.22
J66268.06EA 46.6 EUR
J66268.06 J66268.14 J66268.22
Sulphadoxin 98%
Sulfadoxin

Purity: 98% CAS No.: 2447-57-6 Molecular Formula: C12H14N4O4S Formula Weight: 310.33 Physical Form: Powder Appearance: White Solubility: Soluble in ethanol & ammonium hydroxide (9:1) at 20mg/ml Beilstein Reference: 625453 MDL No.: MFCD00792890

Formel: C₁₂H₁₄N₄O₄S
Molecular Weight: 310,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00792890
CAS-Nummer: 2447-57-6
EINECS: 219-504-9
Merck Index: 14,8906

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR