Sparfloxacin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Amino-1-cyclopropyl-7-((3S,5R)-3,5-dimethylpiperazin-1-yl)-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Purity: 98%
CAS No.: 110871-86-8
Molecular Formula: C19H22F2N4O3
Formula Weight: 392.40
Storage Temperature: 2°C to 8°C
Flash Point:
Applications: Targets a wide range of gram positive, gram negative, and mycoplasma species
Formel:
C₁₉H₂₂F₂N₄O₃ Molecular Weight: 392,407 g/mol Siedepunkt: 640 °C Schmelzpunkt: 265 °C Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00869619 CAS-Nummer: 110871-86-8 Merck Index: 14,8735 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...